Learn More
ABCA1 Antibody (5A1-1422.11) [Janelia Fluor™ 635], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB400-165JF635
This item is not returnable.
View return policy
Description
ABCA1 Monoclonal antibody specifically detects ABCA1 in Mouse samples. It is validated for Flow Cytometry,Western Blot (Negative)
Specifications
| ABCA1 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| ABC-1, ABC1ATP-binding cassette transporter A1, ATP-binding cassette 1, ATP-binding cassette transporter 1, ATP-binding cassette, sub-family A (ABC1), member 1, CERP, Cholesterol efflux regulatory protein, EC 2.7.7.8, EC 2.8.1.8, EC 3.6.3, EC 3.6.3.41, FLJ14958, HDLDT1, membrane-bound, MGC164864, MGC165011, TGDATP-binding cassette sub-family A member 1 | |
| Mouse ABCA1 Antibody (5A1-1422.11) expressing HeLa stable transfectants. [UniProt# P41233] | |
| 0.1 mL | |
| ABC Transporters, Astrocyte Markers, Cancer, Cardiovascular Biology, Cellular Markers, Cholesterol Metabolism, Diabetes Research, Glia Markers, Golgi Apparatus Markers, Lipid and Metabolism, Neuroscience, Signal Transduction | |
| 19 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Flow Cytometry, Western Blot (Negative) | |
| 5A1-1422.11 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |