missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AARSD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14900-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
AARSD1 functions in trans to edit the amino acid moiety from incorrectly charged tRNA(Ala).Specifications
AARSD1 | |
Polyclonal | |
Unconjugated | |
AARSD1 | |
AARSD1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
619440, 69684, 80755 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3THG9, Q5XI97, Q9BTE6 | |
Aarsd1 | |
AARSD1 Fusion Protein Ag6692 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |