missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AAGAB Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27319-1-AP-150UL
This item is not returnable.
View return policy
Description
The predicted MW of this protein is 35 kDa. 27319-1-AP recognizes a 37-40 kDa band due to phosphorylation.
The protein encoded by this gene interacts with the gamma-adaptin and alpha-adaptin subunits of complexes involved in clathrin-coated vesicle trafficking. Mutations in this gene are associated with type I punctate palmoplantar keratoderma. Alternatively spliced transcript variants have been found for this gene.Specifications
| AAGAB | |
| Polyclonal | |
| Unconjugated | |
| AAGAB | |
| AAGAB, FLJ11506 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 66939, 79719 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6PD74, Q8R2R3 | |
| Aagab | |
| AAGAB Fusion Protein Ag26340 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction