Learn More
MedChemExpress AA-dUTP sodium salt
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| CAS | 936327-10-5 |
| Product Type | Biochemical Assay Reagents |
| Quantity | 1 mg |
| Shipping Condition | Room temperature |
| Storage Requirements | Please store the product under the recommended conditions in the Certificate of Analysis. |
| Synonym | Aminoallyl-dUTP sodium salt |
| Target | HBV |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.