Learn More
Honeywell Safety Products™ A700 Grey TSR Glasses
Description
- General use and protection against impact (45m/s).
- Well suited for indoor/outdoor applications such as utilities and light manufacturing.
- Affordably priced and economical.
- Sporty styling.
- Secure and comfortable fit.
- Lightweight design.
- Soft temple tip pads.
- Molded universal nose bridge.
- 8-base polycarbonate, abrasion resistant lenses.
- 180° field of distortion-free vision.
Specifications
Specifications
| Certifications/Compliance | EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number EC1549, EC Attestation Number EC1549 |
| Lens Coating | Anti-fog, Impact-resistant |
| Lens Tint | Gray |
| Product Type | Safety Glasses |
| For Use With (Application) | General use and protection against impact (45m/s). Well suited for indoor / outdoor applications such as utilities and light manufacturing. Building, Construction, Catering, Food, Logistics, Maintenance, Service, Telecom, Transportation and Utility industries. |
| Frame Color | Gray |
| No. per Pack | 1 Ea. |
| Lens Material | Polycarbonate |
| Frame Material | Polycarbonate |