Learn More
Brady™ A4 Signs Set
Click to view available options
Height (Metric):
297 mm
Unit Size:
Each
Specifications
Specifications
| Quantity | 1 Ea. |
| Height (Metric) | 297 mm |
| Width (Metric) | 210 mm |
| Dimensions (L x W) | 210 x 297 mm |
| Product Type | A4 Sign Set |
| Color | Black |