missing translation for 'onlineSavingsMsg'
Learn More

A 83-01, MedChemExpress

Product Code. p-200097200
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16463968

Brand: Medchem Express HY10432

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress A 83-01 is a potent inhibitor of TGF-β type I receptor ALK5 kinase, type I nodal receptor ALK4 and type I nodal receptor ALK7, with IC50s of 12 nM, 45 nM and 7.5 nM against the transcription induced by ALK5, ALK4 and ALK7, respectively.

A 83-01 is a potent inhibitor of TGF-β type I receptor ALK5 kinase, type I nodal receptor ALK4 and type I nodal receptor ALK7, with IC50s of 12 nM, 45 nM and 7.5 nM against the transcription induced by ALK5, ALK4 and ALK7, respectively.

Specifications

Chemical Name or Material A 83-01
CAS 909910-43-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C25H19N5S
Quantity 10 mg
Solubility Information DMSO : 25 mg/mL (59.31 mM; ultrasonic and warming and heat to 60°C) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES S=C(N1N=C(C2=NC(C)=CC=C2)C(C3=CC=NC4=CC=CC=C34)=C1)NC5=CC=CC=C5
Molecular Weight (g/mol) 421.52
Formula Weight 421.52
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.