Description
Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection. Outstanding resistance to heat, UV, and ozone. Use in temperatures ranging from -60°C to 135°C (-75°F to 275°F). High flexural fatigue strength—ideal for use in peristaltic pumps. Heat sealable—no fittings needed to join tubing
Specifications
Specifications
| Diameter (English) Inner | 1/4 in. |
| Material | Tygon A-60-G |
| Diameter (English) Outer | 1/2 in. |
| Overall Length (Metric) | 50 ft. |
| Color | Black |