missing translation for 'onlineSavingsMsg'
Learn More

A-443654, MedChemExpress

Product Code. p-200100396
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16433968

Brand: Medchem Express HY10425

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress A-443654 is a pan-Akt inhibitor and has equal potency against Akt1, Akt2, or Akt3 within cells (Ki=160 pM).

A-443654 is a pan-Akt inhibitor and has equal potency against Akt1, Akt2, or Akt3 within cells (Ki=160 pM).

Specifications

Chemical Name or Material A-443654
CAS 552325-16-3
Color White
Purity Grade Notes Research
Molecular Formula C24H23N5O
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (251.59 mM)
SMILES N[C@H](COC1=CN=CC(C2=CC3=C(NN=C3C)C=C2)=C1)CC4=CNC5=CC=CC=C54
Molecular Weight (g/mol) 397.47
Formula Weight 397.47
Percent Purity 98.5%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.