Learn More
A-443654, MedChemExpress
Product Code. 16433968
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16433968 10 mg 10mg
16416687 5 mg 5mg
16401059 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16433968 Supplier Medchem Express Supplier No. HY10425
MedChemExpress A-443654 is a pan-Akt inhibitor and has equal potency against Akt1, Akt2, or Akt3 within cells (Ki=160 pM).
A-443654 is a pan-Akt inhibitor and has equal potency against Akt1, Akt2, or Akt3 within cells (Ki=160 pM).
Specifications
Chemical Name or Material A-443654
CAS 552325-16-3
Color White
Purity Grade Notes Research
Molecular Formula C24H23N5O
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (251.59 mM)
SMILES N[C@H](COC1=CN=CC(C2=CC3=C(NN=C3C)C=C2)=C1)CC4=CNC5=CC=CC=C54
Molecular Weight (g/mol) 397.47
Formula Weight 397.47
Percent Purity 98.5%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease