Learn More
Thermo Scientific™ 9mm Autosampler Inserts
Product Code. 12624045 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
325 μL
375 μL
400 μL
500 μL
Material:
Glass
Molded Plastic
Polypropylene
Product Type:
Autosampler
Autosampler Vial Insert
Base Style:
Conical
Conical, Silanized
Flat Bottom
Flat Bottom, Silanized
Precision Point
Pulled Point
Unit Size:
Pack of 100
Pack of 500
Product Code. Volume (Metric) Material Product Type Base Style unitSize
12624045 500 μL Molded Plastic Autosampler Vial Insert Flat Bottom Pack of 500
11911563 400 μL Glass Autosampler Vial Insert Pulled Point Pack of 100
12403668 375 μL Glass Autosampler Vial Insert Conical Pack of 100
12812344 400 μL Glass Autosampler Vial Insert Precision Point Pack of 100
12423668 325 μL Polypropylene Autosampler Vial Insert Conical Pack of 100
11961563 375 μL Glass Autosampler Vial Insert Conical Pack of 100
11941533 500 μL Glass Autosampler Vial Insert Flat Bottom Pack of 500
12493708 375 μL Glass Autosampler Vial Insert Conical, Silanized Pack of 100
12403718 375 μL Glass Autosampler Vial Insert Conical Pack of 100
12410078 500 μL Glass Autosampler Flat Bottom, Silanized Pack of 500
12443708 375 μL Glass Autosampler Vial Insert Conical Pack of 100
12133771 325 μL Polypropylene Autosampler Vial Insert Conical Pack of 100
12 options
Product Code. 12624045
Brand: Thermo Scientific™ C4011631P
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Clear glass or polypropylene inserts for 9mm HPLC and GC autosampler vials are available in a variety of configurations with usable volumes from 160μL to 3.5mL.
  • Pulled point inserts are an economical choice for noncritical applications
  • Nominal O.D. of 6mm to fit Target wide-opening vials
For use with:
  • Target LoVial Wide-Opening vials
  • Snap-It™ Wide-Opening Snap Cap vials
  • Target DP™ vials
  • Target Wide-Opening vials
TRUSTED_SUSTAINABILITY
Specifications
Description 400µL MicroSert Insert
Diameter (Metric) Inner 6 mm
Color Clear
Diameter (Metric) Outer 6 mm
Material Molded Plastic
Volume (Metric) 500 μL
Insert Type MicroSert Insert
Volume (Metric) Residual <25 μL
Base Style Flat Bottom
Volume (Metric) Total 500 μL
Quantity 500
Volume (Metric) Usable 400 μL
For Use With (Equipment) Target DP Vials
Certifications/Compliance Certificate of Analysis (COA), Certificate of Conformance (COC)
Height (Metric) 31 mm
Product Type Autosampler Vial Insert