missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 99.9% Isopropanol, Molecular Biology Grade

Product Code. 30117589 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Safe-Cote™ Amber Glass 
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
30117589 1 L Safe-Cote™ Amber Glass  1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30117589 Supplier Thermo Scientific™ Supplier No. T036181000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Prequalified for molecular biology applications | C3 H8 O | 60.1 g/mol

Suitable for the purification and precipitation of nucleic acids (DNA and RNA) and proteins

  • 99.9% Isopropanol
  • DNase-, RNase-, and protease-free
  • Pre-qualified for molecular biology applications
  • Suitable for the purification and precipitation of nucleic acids (DNA and RNA) and proteins
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isopropyl alcohol
pH 7, 1% aq. sol
Melting Point -89.5°C
Absorbance <=1.00 @ 205 NM, <=0.20 @ 220 NM, <=0.10 @ 230 NM, <=0.015 @ 254 NM
Color Colorless
Appearance Clear, Colorless Liquid
Quantity 1 L
CAS 67-63-0
Packaging Safe-Cote™ Amber Glass 
Residue after Evaporation <=1 ppm
Molecular Formula C3 H8 O
Water <=0.05%
Synonym 2-Propanol; IPA; Isopropyl alcohol; Propan-2-ol; Isopropanol
Boiling Point 81°C to 83°C
Molecular Weight (g/mol) 60.1
Percent Purity ≥99.5%
Grade Molecular Biology
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.