Learn More
SPEX™ 96-Well Titer Plate with Round Wells
Product Code. 13561190 Shop All SPEX Certiprep Products
Change view
Click to view available options
Capacity (Metric):
96 x 1 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
13561190 96 x 1 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13561190 Supplier SPEX™ Supplier No. 2210SPP
SPEX™ 96-Well Titer Plate features round wells constructed of durable polypropylene.
  • Extensively tested with various samples and shown to resist perforation by grinding media
  • Compatible with 2650 Cryo-Adapter to preserve temperature sensitive samples
  • Alphanumeric marks for well identification
  • Durable polypropylene construction
  • Process 192 samples at a time by running two titer plates in the Standard Clamp Assembly
  • Process 384 samples at a time by running four titer plates in the Large Capacity Clamp Assembly
  • Deep-well titer plate with a 96-well format
  • Each round-bottom well has a volume of 1.0 mL and a working volume of 0.7 mL
  • To use, simply load each well with the sample, solution (if desired), and grinding media, then seal with a 2211 Cap-Mat. Place up to six titer plates in the Geno/Grinder™ for high-throughput tissue homogenization and cell lysis
Specifications
Product Type 96-Well Plate
Capacity (Metric) 96 x 1 mL