Learn More
uvex 95798 Insole For Uvex 1 Sport - Width 11
Product Code. 17298971 Shop All Uvex Products
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
49
51
Unit Size:
1 pair
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17298971 40 1 pair
17248971 35 1 pair
17258971 36 1 pair
17268971 37 1 pair
17278971 38 1 pair
17288971 39 1 pair
17208981 41 1 pair
17218981 42 1 pair
17228981 43 1 pair
17238981 44 1 pair
17248981 45 1 pair
17258981 46 1 pair
17268981 47 1 pair
17278981 48 1 pair
17288981 49 1 pair
17298981 51 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Product Code. 17298971 Supplier uvex Supplier No. 9579840
Only null left
Add to basket
Add to basket
Comfortable climatic insole with antistatic properties for Uvex 1 Sport
  • Full-surface foot shock absorption
  • Highly breathable and moisture-absorbent material
  • Shock absorption zones support natural foot movement
  • Skin compatibility and pleasant surface structure
  • Supports foot arch
  • Article No. 9579.8
  • Sizes: 35 to 52
TRUSTED_SUSTAINABILITY
Specifications
Color Black
For Use With (Equipment) Uvex 1 Sport
Material Microvelour
Size 40
Disposable Single-use
Gender Unisex
Fit Standard Width
Latex Free Yes
Waterproof No
Product Type Insoles