Learn More
Uvex™ 9197 Skyper SX2 Safety Spectacles
Click to view available options
Supravision Excellence
Description
- Extremely lightweight and modern safety spectacle with outstanding field of vision and exceptional functionality
- Supravision excellence coating is anti-fog on the inside, and extremely scratch-resistant and chemical resistant on the outside Supravision sapphire coating is extremely scratch-resistant and chemical-resistant on both sides
- Individual temple inclination and length adjustment for a personalized fit
- Soft temples
- Simple lens replacement
Specifications
Lens Type | Supravision Excellence |
Certifications/Compliance (Europe) | EN 166:2001, EN 172:1994 + A1:2000 + A2:2001 |
Lens Tint | Clear |
Frame Color | Blue |
Model | Skyper SX2 |
Lens Material | Polycarbonate |
UV Protection | UV 400 |