Learn More
Uvex™ 9197 Skyper SX2 Safety Spectacles
Catalog Number 10778153
Click to view available options
Supravision Excellence
Successful symbiosis of maximum functionality, optimal safety, and fashionable appearance.
  • Extremely lightweight and modern safety spectacle with outstanding field of vision and exceptional functionality
  • Supravision excellence coating is anti-fog on the inside, and extremely scratch-resistant and chemical resistant on the outside
  • Supravision sapphire coating is extremely scratch-resistant and chemical-resistant on both sides
  • Individual temple inclination and length adjustment for a personalized fit
  • Soft temples
  • Simple lens replacement
Lens Type Supravision Excellence
Certifications/Compliance (Europe) EN 166:2001, EN 172:1994 + A1:2000 + A2:2001
Lens Tint Clear
Frame Color Blue
Model Skyper SX2
Lens Material Polycarbonate
UV Protection UV 400