Learn More
Uvex™ 9188 Cybric Safety Spectacles
167.06 NOK valid until 2024-04-30
Use promo code "21673" to get your promotional price.
Use promo code "21673" to get your promotional price.
Click to view available options
Lens Tint:
Clear
Gray
Description
- Spectacles feature a large field of vision and individual adjustment capabilities for reliable protection and comfort
- Certified according to EN 166 (personal eye protection) and EN 170 (UV filters)
- Quattroflex side arms with four comfort cushions to ensure a comfortable, pressure-free fit around the ear
- Inclination of the side arms can be adjusted between multiple levels, allowing them to be adapted individually to all facial shapes
- Supravision sapphire coating is extremely chemical-resistant and scratch-resistant on both sides
- Reliable UV 400 protection
Specifications
Lens Type | Supravision Sapphire |
Certifications/Compliance (Europe) | EN 166:2001, EN 172:1994 + A1:2000 + A2:2001 |
Lens Tint | Gray |
Frame Color | Black, Orange |
Model | 9188 Cybric |
Lens Material | Polycarbonate |
UV Protection | UV 400 |