Learn More
uvex i-5 Safety Glasses with CBR65 Lens
Reliable protection with an innovative, modern design and the x-tended eyeshield and sideshield. The uvex i-5 can be relied on to protect the eyes in many situations.
201.00 NOK - 215.00 NOK
Specifications
| Lens Coating | Anti-fog |
|---|---|
| Lens Type | Supravision Excellence |
| Product Type | Ergonomic Safety Glasses |
| Model | 9183 i-5 |
| Material | Polycarbonate |
| Product Code | Brand | Lens Tint | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Lens Tint | Price | Quantity & Availability | |||||
|
17207271
|
uvex
9183265 |
Clear |
201.00 NOK
Pack of 1 |
Estimated Shipment: 26-05-2026
Log in to see stock. |
|||||
|
17217271
|
uvex
9183281 |
Gray |
215.00 NOK
Pack of 1 |
Estimated Shipment: 10-06-2026
Log in to see stock. |
|||||
Description
- Sporty premium safety glasses with a variety of features and customization options
- Robust polycarbonate lens with proven uvex supravision coating technology – permanently anti-fog and scratch-resistant lens
- Optional extra frame provides greater protection against particles, dust and dirt
- Metal-free
- Frame color: white, black
- PC lens: CBR65 (light brown)
- Certified in line with EN 166 (personal eye protection) and EN 172 (sunglare filters for industrial use)
- Marking: W 166 FT CE – 5-1.4 W1 FTKN CE
- UV400 – 100% protection against dangerous UVA and UVB radiation to 400 nm
- X-tended eyeshield: soft-seal frame technology wraps the forehead and provides protection against particle penetration
- X-tended sideshield offers additional side protection
- Ergonomically shaped frame with multi-stage adjustable side arms allows a high degree of customization in order to guarantee a good fit on any head or face shape
- Soft ear pieces for a comfortable, secure hold without creating pressure points
- Soft, flexible nose pad adapts to the wearer and ensures a comfortable, pressure-free fit
Specifications
| Anti-fog | |
| Ergonomic Safety Glasses | |
| Polycarbonate | |
| Polycarbonate | |
| UV400 |
| Supravision Excellence | |
| 9183 i-5 | |
| Reusable | |
| Plastic | |
| Standard |