Learn More
Uvex™ 9172 Super G Safety Glasses
Product Code. 15305922 Shop All Uvex Products
Click to view available options
:
Supravision Sapphire
Product Code. 15305922
Brand: Uvex™ 9172085
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The lightest safety glasses in the world weighing only 18 grams with full safety, modern design and an innovative temple concept.
  • The lightest safety glasses in the world weighing only 18 grams with full safety, modern design and an innovative temple concept
  • Supravision excellence coating is anti-fog on the inside, and extremely scratch-resistant and chemical-resistant on the outside
  • Supravision plus coating is anti-fog and scratch-resistant on both sides
  • Supravision sapphire coating is extremely scratch-resistant and chemical-resistant on both sides
  • Aerodynamically optimized and inclined lens geometry
  • Soft temple ends for maximum wearing comfort
  • Easy lens replacement
TRUSTED_SUSTAINABILITY
Specifications
Lens Type Supravision Sapphire
Certifications/Compliance (Europe) EN 166:2001, EN 170:2002
Lens Tint Clear
Frame Color Gray
Disposable Reusable
Lens Material Polycarbonate
UV Protection UV 400
Autoclavable Not Autoclavable