Learn More
Uvex™ 9169 Super F OTG Welding Safety Spectacles
Catalog Number 17227261
Click to view available options
Infradur Plus, Gray
These extremely comfortable safety spectacles provide excellent eye area protection, and fit perfectly over conventional prescription lenses. Lens coating provides protection against burns caused by welding sparks.
  • Overspecs for users who also wear prescription lenses with an arm-hinge concept
  • Certified in line with EN 166 (personal eye protection) and EN 169 (welding protection)
  • Side arms graduate to soft, flexible material for enhanced comfort and grip
  • Spectacles are available in a variety of welding protection levels
  • Sunglare filter allows for perfect color recognition
  • Unrestricted peripheral vision
Infradur Plus Coating
  • Inside: Anti-fog
  • Outside: Scratch-resistant
  • Minimizes burns due to welding sparks
Lens Type Infradur Plus
Description Welding Protection Level: 1.7
Certifications/Compliance (Europe) EN 166:2001, EN 169:2001
Lens Tint Black
Frame Color Green
UV Protection UV 400
Labeling W 166 FT CE - 1,7 W 1 FTKN CE