missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 9-Isopropylcarbazole, 97%

Catalog Number 11416610
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
1484-09-9
C15H15N
209.29
MFCD00046896
LSZJZNNASZFXKN-UHFFFAOYSA-N
9-isopropylcarbazole, 9-isopropyl-9h-carbazole, carbazole, 9-isopropyl, 9h-carbazole, 9-1-methylethyl, n-isopropylcarbazole, 9-isopropyl carbazole, 9-propan-2-yl-9h-carbazole, 9-methylethyl carbazole, maybridge3_000552, acmc-1c0tf
73873
9-propan-2-ylcarbazole
CC(C)N1C2=C(C=CC=C2)C2=C1C=CC=C2
This item is not returnable. View return policy
1484-09-9
C15H15N
209.29
MFCD00046896
LSZJZNNASZFXKN-UHFFFAOYSA-N
9-isopropylcarbazole, 9-isopropyl-9h-carbazole, carbazole, 9-isopropyl, 9h-carbazole, 9-1-methylethyl, n-isopropylcarbazole, 9-isopropyl carbazole, 9-propan-2-yl-9h-carbazole, 9-methylethyl carbazole, maybridge3_000552, acmc-1c0tf
73873
9-propan-2-ylcarbazole
CC(C)N1C2=C(C=CC=C2)C2=C1C=CC=C2
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'