missing translation for 'onlineSavingsMsg'
Learn More

9-beta-D-Arabinofuranosylguanine, Thermo Scientific™

Product Code. p-7232411 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. Quantity unitSize
17332491 100 mg 100mg
17342491 500 mg 500mg
2 options
This item is not returnable. View return policy

Product Code. 17332491

Brand: Thermo Scientific Acros 468631000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 38819-10-2
Molecular Formula C10H13N5O5
Molecular Weight (g/mol) 283.24
MDL Number MFCD00065486
InChI Key NYHBQMYGNKIUIF-FJFJXFQQSA-N
Synonym Araguanosine; Ara-G
IUPAC Name 2-amino-9-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-3H-purin-6-one
SMILES NC1=NC(=O)C2=C(N1)N(C=N2)[C@@H]1O[C@H](CO)[C@@H](O)[C@@H]1O

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Physical Form Crystalline Powder
Color White
Melting Point 225°C
Quantity 100 mg
Formula Weight 283.24
Chemical Name or Material 9-beta-D-Arabinofuranosylguanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.