Learn More
9-[4-Acetoxy-3-(hydroxymethyl)but-1-yl]-2-aminopurine, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 104227-88-5 |
| Molecular Formula | C12H17N5O3 |
| MDL Number | MFCD08063728 |
| Quantity | 100 mg |
| SMILES | CC(OCC(CO)CCN1C=NC2=CN=C(N)N=C12)=O |
| Molecular Weight (g/mol) | 279.30 |
| Percent Purity | 95% |
| Chemical Name or Material | 9-[4-Acetoxy-3-(hydroxymethyl)but-1-yl]-2-aminopurine |