Learn More
9-[2-(Diethylphosphonomethoxy)propyl-d6] Adenine, TRC
Product Code. 30372610
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30372610 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30372610 Supplier Toronto Research Chemicals Supplier No. TRCD4447521MG
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 9-[2-(Diethylphosphonomethoxy)propyl-d6] Adenine
CAS 1020719-38-3
Molecular Formula C13 2H6 H16 N5 O4 P
Quantity 1 mg
InChI Formula InChI=1S/C13H22N5O4P/c1-4-21-23(19,22-5-2)9-20-10(3)6-18-8-17-11-12(14)15-7-16-13(11)18/h7-8,10H,4-6,9H2,1-3H3,(H2,14,15,16)/i6D2,7D,8D,9D2
SMILES [2H]c1nc(N)c2nc([2H])n(c2n1)C([2H])([2H])C(C)OC([2H])([2H])P(=O)(OCC)OCC
IUPAC Name 2,8-dideuterio-9-[1,1-dideuterio-2-[dideuterio(diethoxyphosphoryl)methoxy]propyl]purin-6-amine
Molecular Weight (g/mol) 349.36
Formula Weight 349.1786