Learn More
Fisherbrand™ 8mm PP Screw Seal, Black, Closed top, 8-425 thread, Assembled septum
Shop All Fisherbrand Products
Click to view available options
Color:
Beige, Red-Orange, Black
Blue Transparent, White, Black
Cream, Red, Black
Gray, Red, Black
Red, White, Black
Red-Orange, Transparent, Black
Material (Liner):
Natural Rubber/TEF
PTFE/Butyl
Red Rubber/PTFE
Silicone/PTFE
Thickness (Metric):
1 mm
1.3 mm
1.5 mm
Unit Size:
Pack of 100
Pack of 1000
Description
- 8mm
- PP
- Screw Cap
- Black
- Closed top
- 8-425 thread
- Pre-assembled caps and septa are convenient and minimize contamination from handling
Specifications
Specifications
| Material (Liner) | Red Rubber/PTFE |
| Color | Beige, Red-Orange, Black |
| Diameter (Metric) Outer | 8 mm |
| Material | Polypropylene |
| For Use With (Application) | GC, HPLC |
| Quantity | 100 |
| Thickness (Metric) | 1 mm |