Learn More
Cytiva 850-DS PVDF 8-Channel Filter Plates
Used for filtering purposes.
Brand: Cytiva 7707-3800
6089.50 NOK valid until 2024-12-31
Use promo code "23266" to get your promotional price.
Additional Details : Weight : 1.70000kg
Description
- Used for filtering purposes/li>
Specifications
PVDF | |
Filtering | |
850-DS PVDF 8-Channel Filter Plates |
0.45 μm | |
50/Pk. |