Learn More
Thermo Scientific™ 81-Well Cryogenic Storage Racks with Lid
GREENER_CHOICE
Product Code. p-4520568 Shop All MBP Molecular BioProducts Products
Change view
Click to view available options
Color:
Fluorescent Green
Fluorescent Orange
Fluorescent Pink
Natural
Unit Size:
Each
Pack of 20
Product Code. Color unitSize
11570115 Natural Pack of 20
10461411 Fluorescent Green Each
10781592 Fluorescent Orange Pack of 20
12358192 Fluorescent Pink Each
4 options
Product Code. 11570115
Brand: Thermo Scientific™ 8800
Only null left
Add to basket
Add to basket
Hold eight-one 1.5 or 2.0mL cryogenic storage vials with this stackable cryogenic storage rack with lid.
Hold eighty-one 1.5 or 2.0mL cryogenic storage vials with the Thermo Scientific™ 81-Well Cryogenic Storage Rack with Lid. Available in several colors with transparent tinted lid that protects samples from dust and debris and permits stacking.
  • Molded with alphanumeric grid for easy tube identification
  • Lid protects samples and allows racks to be stacked
TRUSTED_SUSTAINABILITY
Specifications
Color Natural
Holds 81 (1.5 or 2.0 mL Tubes)
Material Polypropylene
Type 81-Well Cryogenic Storage Racks with Lid