Learn More
Thermo Scientific™ 8 x 50 Individually Sealed Fixed Angle Rotor for 50 mL conical (for compact and 1.6L General Purpose Centrifuges)
Product Code. 10605835 Shop All Thermo Scientific Products
Click to view available options
Capacity:
8 x 50 mL
Unit Size:
Each
Product Code. 10605835
Brand: Thermo Scientific™ 75003694
Centrifuge eight conical tubes in individually sealed vessels.
Centrifuge eight conical tubes in individually sealed vessels using Thermo Scientific™ 8 x 50mL Sealed Fixed-Angle Rotor.
Individual sealed canisters for high-risk samples, certified by Public Health England, Porton Down, UK.
Secure, push-button Auto-Lock rotor exchange in as little as 3 seconds delivers:
  • Trouble-free rotor installation and removal
  • No tools are required
  • No complicated rotor changing procedures
  • Clear chamber access for cleaning convenience
  • Quick switch between applications, from conical tubes to blood tubes, from microtubes to microplates, and the flexibility to evolve with the changing needs of your laboratory
TRUSTED_SUSTAINABILITY
Specifications
Description 8 x 50mL Individually Sealed Fixed Angle Rotor
Type Fixed Angle Rotor
Capacity 8 x 50 mL
Max. RCF 6,755 x g
Max. Speed 6,500 rpm
Radius (Metric) 143 mm
Max. Tube Length 116 mm
Max. Tube Diameter 29.5 mm
For Use With (Equipment) Thermo Scientific Sorvall Legend X1, Sorvall ST 16, Multifuge X1, Megafuge 16 and SL 16 Centrifuges
Angle 25°