Learn More
8-Octadecyloxypyrene-1,3,6-trisulfonic Acid Trisodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | 8-Octadecyloxypyrene-1,3,6-trisulfonic Acid Trisodium Salt |
| CAS | 111024-84-1 |
| Molecular Formula | C34 H43 Na3 O10 S3 |
| Quantity | 1 mg |
| InChI Formula | InChI=1S/C34H46O10S3.3Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-22-44-29-23-30(45(35,36)37)26-20-21-28-32(47(41,42)43)24-31(46(38,39)40)27-19-18-25(29)33(26)34(27)28;;;/h18-21,23-24H,2-17,22H2,1H3,(H,35,36,37)(H,38,39,40)(H,41,42,43);;;/q;3*+1/p-3 |
| SMILES | CCCCCCCCCCCCCCCCCCOc1cc(c2ccc3c(cc(c4ccc1c2c34)S(=O)(=O)O[Na])S(=O)(=O)O[Na])S(=O)(=O)O[Na] |
| IUPAC Name | [3-octadecoxy-6,8-bis(sodiooxysulfonyl)pyren-1-yl]sulfonyloxysodium |
| Molecular Weight (g/mol) | 776.864 |
| Formula Weight | 776.171 |