missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Methyl Varenicline Dihydrochloride, TRC
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
Specifications
Specifications
| Chemical Name or Material | 8-Methyl Varenicline Dihydrochloride |
| Molecular Formula | C14H15N3 • 2HCl |
| Quantity | 500 mg |
| Synonym | 8-Methyl-7,8,9,10-tetrahydro-6H-6,10-methanoazepino[4,5-g]quinoxaline Dihydrochloride |
| InChI Formula | InChI=1S/C14H15N3.2ClH/c1-17-7-9-4-10(8-17)12-6-14-13(5-11(9)12)15-2-3-16-14;;/h2-3,5-6,9-10H,4,7-8H2,1H3;2*1H |
| SMILES | CN1CC2CC(C3=CC4=NC=CN=C4C=C23)C1.Cl.Cl |
| IUPAC Name | 8-methyl-7,8,9,10-tetrahydro-6H-6,10-methanoazepino[4,5-g]quinoxaline dihydrochloride |
| Molecular Weight (g/mol) | 298.21 |
| Formula Weight | 297.08 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction