Learn More
Cytiva Whatman™ 8-Channel Filter Plates
For use with Agilent 850-DS dissolution sampling station
Brand: Cytiva 7707-3200
6089.50 NOK valid until 2024-12-31
Use promo code "23266" to get your promotional price.
Additional Details : Weight : 1.71000kg
Description
- 0.45μm membrane pore size
- Available in PTFE, Nylon, PES, and GMF filter media
- Disposable
Automated dissolution testing
Specifications
Agilent 850-DS Dissolution Sampling Station | |
PES | |
0.45 μm | |
Automated dissolution testing |