Learn More
Integra Biosciences™ 8-channel Adapter
Product Code. 12603275
Change view
Click to view available options
Type:
8-Channel adapter with ejector for grip tips
8-Channel stainless steel adapter, 40mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
12603275 8-Channel adapter with ejector for grip tips Each
10023451 8-Channel stainless steel adapter, 40mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12603275 Supplier Integra Biosciences™ Supplier No. 159025
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
Estimated Shipment: 02-11-2026
Use these adapters with VACUBOY aspiration tools. Integra Biosciences™ 8-Channel Adapters used with VACUBOY aspiration tools convert any vacuum source into a user-friendly aspiration system.
  • 8 channel plastic aspiration adapter for disposable tips with ejector
  • Fit a variety of tubes, flasks, bottles etc.
  • Accommodate 12- to 96-well plates
Can be used with VACUSAFE comfort vacuum system. Enables liquid residues to be disposed of reliably, precisely and conveniently:
  • With a precisely dosed volume
  • Without loss of drops
  • Without risk of contamination
  • Without causing tiredness
  • All parts in contact with fluids are autoclavable
Specifications
Type 8-Channel adapter with ejector for grip tips
For Use With (Equipment) VACUBOY Aspiration Tools