Learn More
8-Benzyloxy-5-(2-bromoacetyl)-2(1H)-quinolinone, TRC
Product Code. 30337251
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30337251 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30337251 Supplier Toronto Research Chemicals Supplier No. TRCB286115100MG
 Restricted Item
Estimated Shipment: 08-10-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 8-Benzyloxy-5-(2-bromoacetyl)-2(1H)-quinolinone
CAS 100331-89-3
Molecular Formula C18H14BrNO3
Quantity 100 mg
Synonym 5-(Bromoacetyl)-8-(phenylmethoxy)-2(1H)-quinolinone, 5-(2-Bromo-1-oxoethyl)-8-benzyloxy-2(1H)-quinolinone, 5-Bromoacetyl-8-benzyloxycarbostyril, 8-(Benzyloxy)-5-(bromoacetyl)quinolin-2(1H)-one, 8-Benzyloxy-5-(2-Bromoacetyl)-1H-quinolin-2-one, 8-Benzyloxy-5-bromoacetylcarbostyril
InChI Formula InChI=1S/C18H14BrNO3/c19-10-15(21)13-6-8-16(18-14(13)7-9-17(22)20-18)23-11-12-4-2-1-3-5-12/h1-9H,10-11H2,(H,20,22)
SMILES BrCC(=O)c1ccc(OCc2ccccc2)c3NC(=O)C=Cc13
IUPAC Name 5-(2-bromoacetyl)-8-phenylmethoxy-1H-quinolin-2-one
Molecular Weight (g/mol) 372.21
Formula Weight 371.0157