Learn More
MP Biomedicals™ 7X™-O-Matic Solution
Product Code. 11439585 Shop All MP Biomedicals Products
Change view
Click to view available options
Volume (English):
1 gal.
5 gal.
4 x 1 gal.
Unit Size:
Each
Pack of 4
Product Code. Volume (English) unitSize
11439585 1 gal. Each
11449585 4 x 1 gal. Pack of 4
11459585 5 gal. Each
3 options
Product Code. 11439585
Brand: MP Biomedicals™ 097667493
7X™-O-Matic Solution was developed for safe and effective cleaning of laboratory instruments and glassware under pressure, heat and agitation (machine wash)
  • Developed for safe and effective cleaning of laboratory instruments and glassware
  • Formulated to be used in a machine washer to withstand heat, pressure and agitation
  • Powerful wetting and penetrating agent, which contains a non-ionic surfactant and a hydrotrope to complex phosphates
  • Behaves and remains mostly suds-free when agitated
  • Several convenient package sizes are available, and it is stable for long-term storage
  • Effective and safe
  • Eliminates residue leaving no interfering fluorescence
  • No etch to glass or plastic labware at any concentration
  • Completely drains from glassware in seconds
TRUSTED_SUSTAINABILITY
Specifications
Form Liquid
pH 9 to 11
Appearance Clear to Light Yellow
Concentration 7X
Content And Storage 15°C to 30°C
For Use With (Application) Use on Glass, Plastic, benchtop equipment and tubing
Volume (English) 1 gal.