Learn More
Ansell™ 76-301 Series Tiger Paw™ PVC Light Weight Gloves
Product Code. 12626991 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
Unit Size:
12 pairs
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12626991 7 12 pairs
12616991 10 12 pairs
12636991 8 12 pairs
12646991 9 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Product Code. 12626991 Supplier Ansell™ Supplier No. 76301070
Estimated Shipment: 17-11-2026
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Made from polyester plated with cotton, Tiger Paw™ offers many features and benefits at an economical price. This heavier construction ensures a more robust glove for heavier applications.
  • Made from polyester plated with cotton
  • Knitted construction
  • Polyester cotton liner
  • Dotted on palm side finish
  • Knitwrist cuff
TRUSTED_SUSTAINABILITY
Specifications
Back Material PVC
Certifications/Compliance Standard CATEGORY II EN 388 : 214X
Cuff Material PVC
Color Natural, Blue
Product Type Lightweight Gloves
Size 7
Glove Cuff Style Knit
Lining Polyester Cotton
Palm Type Dotted-palm
Quantity 12 Pairs
Series 76-301