Learn More
Uvex™ 7310 silv-Air Pro FFP3 Breathing Mask
Artikelnummer. 10892041 Alle ansehen Uvex Produkte
Change view
Click to view available options
:
FFP3
Artikelnummer.
10892041 FFP3
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 10892041
Marke: Uvex™ 8707310
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Mask provides protection against pollutants up to 30 times the limit value, with an exhalation valve to reduce breathing resistance and heat under the mask.
  • Protection against pollutants up to 30 times the limit value
  • 4-point headband attachment for a comfortable, individual and secure fit of the mask
  • Integrated , flexibly adjustable nose clip
  • Exhalation valve reduces breathing resistance and heat/moisture under the mask
  • Very soft, circumferential sealing lip for optimal wearing comfort and a reliable sealing fit
  • Mask meets the requirements for reusability (R), and against high dust exposure (D marking, dolomite dust test)
TRUSTED_SUSTAINABILITY
Spezifikation
Certifications/Compliance (Europe) EN 149:2001 + A1:2009
Color Gray
With Exhalation Valve Yes
Filter Efficiency FFP3
Model 7310 silv-Air Pro