Learn More
Hamilton™ 700 Series Cemented Needle Syringes
Description
- The original hand-fitted Hamilton™ syringe
- Stainless steel plunger is manufactured to fit the glass barrel with a tolerance less than three thousandths of a millimeter
- Unsurpassed syringe life
Specifications
Specifications
| Material | Borosilicate Glass, Stainless Steel |
| Needle Gauge | 22 G |
| Diameter (Metric) Outer | 7.75 mm |
| Product Type | Syringe |
| Quantity | 1/Ea. |
| Volume (Metric) | 25 μL |
| Needle Point Style | 3 |
| Connection Type | Cemented |