Learn More
Hamilton™ 700 Series Cemented Needle Custom Syringes
Product Code. p-8543004
Change view
Click to view available options
Volume (Metric):
5 μL
10 μL
100 μL
250 μL
Unit Size:
Each
Product Code. Volume (Metric) unitSize
10743903 100 μL Each
10664301 10 μL Each
10359514 250 μL Each
10579672 5 μL Each
4 options
Product Code. 10743903
Brand: Hamilton™ 80608
Syringes for customized needles. This series of syringes is ideal for dispensing volumes from 0.5 up to 500μL.
  • The original hand-fitted Hamilton™ syringe
  • Stainless steel plunger is manufactured to fit the glass barrel with a tolerance less than three thousandths of a millimeter
  • Unsurpassed syringe life
Order Info
This is a Custom Order. Please consult with your Fisher Sales Representative before ordering.
Specifications
Autoclavable Not Autoclavable
Material Borosilicate Glass, Stainless Steel
Needle Gauge Custom
Diameter (Metric) Inner 1.46 mm
Diameter (Metric) Outer 7.75 mm
For Use With (Application) Ideal for dispensing volumes from 0.5 μL up to 500 μL
Product Type Microliter Syringe
Max. Pressure 2000 psig (137.9 bar)
Max. Temperature (Metric) 50°C
Model 710 SN
Min. Temperature (Metric) 10°C
Quantity 1/Ea.
Volume (Metric) 100 μL
Length (Metric) Needle Custom Length
Needle Point Style Custom
Connection Type Cemented Needle Special