Learn More
Hamilton™ 700 Microliter Syringes: N Termination
Product Code. p-4895847
Click to view available options
Volume (Metric):
5 μL
10 μL
25 μL
50 μL
100 μL
250 μL
500 μL
Needle Point Style:
3
Blunt
Cone
Sharp, Bevel, Curved, Non-coring
Diameter (Metric) Inner Needle:
0.13 mm
0.15 mm
0.41 mm
Diameter (Metric) Outer Needle:
0.47 mm
0.483 mm
0.72 mm
Unit Size:
Each
Product Code. 10058300
Brand: Hamilton™ 80765
The 700 series syringe is the original hand-fitted Hamilton syringe. This series of syringes is ideal for dispensing volumes from 0.5 μL up to 500 μL. A variety of different terminations and needle options are available in this series.
  • Original hand-fitted Hamilton syringe
  • Ideal for dispensing volumes from 0.5 µL up to 500 µL
  • A variety of different terminations and needle options are available in this series
Specifications
Autoclavable Not Autoclavable
Material Stainless Steel (Plunger)
Needle Gauge 22 G
Diameter (English) Inner 0.029 in.
Diameter (English) Inner Needle 0.0161 in.
Diameter (Metric) Inner Needle 0.41 mm
Diameter (English) Outer Needle 0.0283 in.
Diameter (English) Outer 0.305 in.
Diameter (Metric) Inner 0.729 mm
Diameter (Metric) Outer 7.75 mm
Diameter (Metric) Outer Needle 0.72 mm
For Use With (Application) For dispensing low volumes to achieve the highest level of accuracy and precision possible
Length (English) 2 in.
Length (English) Needle 2 in.
Length (Metric) 51 mm
Product Type Microliter Syringe
Max. Pressure 2000 psi
Max. Temperature (English) 122°F
Max. Temperature (Metric) 50°C
Min. Temperature (English) 50°F
Model 725
Needle Point Number 3
Quantity Ea.
Volume (Metric) 250 μL
Needle Point Style Blunt
Connection Type Cemented