Learn More
Hamilton™ 700 Microliter Syringes: N Termination
Product Code. 10313811
Change view
Click to view available options
Volume (Metric):
5 μL
10 μL
25 μL
50 μL
100 μL
250 μL
500 μL
Needle Point Style:
3
Blunt
Cone
Sharp, Bevel, Curved, Non-coring
Diameter (Metric) Inner Needle:
0.13 mm
0.15 mm
0.41 mm
Diameter (Metric) Outer Needle:
0.47 mm
0.483 mm
0.72 mm
Unit Size:
Each
Product Code. Volume (Metric) Needle Point Style Diameter (Metric) Inner Needle Diameter (Metric) Outer Needle unitSize
10313811 10 μL Blunt 0.13 mm 0.72 mm Each
10058300 250 μL Blunt 0.41 mm 0.72 mm Each
10095630 500 μL Blunt 0.41 mm 0.72 mm Each
10054414 500 μL Cone 0.41 mm 0.72 mm Each
10097671 10 μL Cone 0.13 mm 0.47 mm Each
10180361 50 μL Blunt 0.15 mm 0.72 mm Each
10778541 25 μL Blunt 0.15 mm 0.72 mm Each
10340711 10 μL 3 0.15 mm 0.72 mm Each
10261821 50 μL Sharp, Bevel, Curved, Non-coring 0.15 mm 0.72 mm Each
10429042 10 μL Sharp, Bevel, Curved, Non-coring 0.13 mm 0.47 mm Each
10149980 25 μL Sharp, Bevel, Curved, Non-coring 0.15 mm 0.72 mm Each
10020332 5 μL 3 0.13 mm 0.483 mm Each
10313571 100 μL Sharp, Bevel, Curved, Non-coring 0.15 mm 0.72 mm Each
10625251 100 μL Blunt 0.15 mm 0.72 mm Each
10251871 250 μL Sharp, Bevel, Curved, Non-coring 0.15 mm 0.72 mm Each
10303621 500 μL Sharp, Bevel, Curved, Non-coring 0.41 mm 0.72 mm Each
16 options
Product Code. 10313811
Brand: Hamilton™ 80383
The 700 series syringe is the original hand-fitted Hamilton syringe. This series of syringes is ideal for dispensing volumes from 0.5 μL up to 500 μL. A variety of different terminations and needle options are available in this series.
  • Original hand-fitted Hamilton syringe
  • Ideal for dispensing volumes from 0.5 µL up to 500 µL
  • A variety of different terminations and needle options are available in this series
Specifications
Autoclavable Not Autoclavable
Material Stainless Steel (Plunger)
Needle Gauge 26s G
Diameter (English) Inner 0.019 in.
Diameter (English) Inner Needle 0.005 in.
Diameter (Metric) Inner Needle 0.13 mm
Diameter (English) Outer Needle 0.0283 in.
Diameter (English) Outer 0.305 in.
Diameter (Metric) Inner 0.485 mm
Diameter (Metric) Outer 7.75 mm
Diameter (Metric) Outer Needle 0.72 mm
For Use With (Application) For dispensing low volumes to achieve the highest level of accuracy and precision possible
Length (English) 2 in.
Length (English) Needle 2 in.
Length (Metric) 51 mm
Product Type Microliter Syringe
Max. Pressure 2000 psi
Max. Temperature (English) 122°F
Max. Temperature (Metric) 50°C
Min. Temperature (English) 50°F
Model 701
Needle Point Number 3
Quantity Ea.
Volume (Metric) 10 μL
Needle Point Style Blunt
Connection Type Cemented