Learn More
Hamilton™ 700 Microliter Syringes: N Termination
Product Code. 10313571
Change view
Click to view available options
Volume (Metric):
5 μL
10 μL
25 μL
50 μL
100 μL
250 μL
500 μL
Needle Point Style:
3
Blunt
Cone
Sharp, Bevel, Curved, Non-coring
Diameter (Metric) Inner Needle:
0.13 mm
0.15 mm
0.41 mm
Diameter (Metric) Outer Needle:
0.47 mm
0.483 mm
0.72 mm
Unit Size:
Each
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Needle Point Style Diameter (Metric) Inner Needle Diameter (Metric) Outer Needle Unit Size
10313571 100 μL Sharp, Bevel, Curved, Non-coring 0.15 mm 0.72 mm Each
10058300 250 μL Blunt 0.41 mm 0.72 mm Each
10095630 500 μL Blunt 0.41 mm 0.72 mm Each
10054414 500 μL Cone 0.41 mm 0.72 mm Each
10097671 10 μL Cone 0.13 mm 0.47 mm Each
10313811 10 μL Blunt 0.13 mm 0.72 mm Each
10180361 50 μL Blunt 0.15 mm 0.72 mm Each
10778541 25 μL Blunt 0.15 mm 0.72 mm Each
10340711 10 μL 3 0.15 mm 0.72 mm Each
10261821 50 μL Sharp, Bevel, Curved, Non-coring 0.15 mm 0.72 mm Each
10429042 10 μL Sharp, Bevel, Curved, Non-coring 0.13 mm 0.47 mm Each
10149980 25 μL Sharp, Bevel, Curved, Non-coring 0.15 mm 0.72 mm Each
10020332 5 μL 3 0.13 mm 0.483 mm Each
10625251 100 μL Blunt 0.15 mm 0.72 mm Each
10251871 250 μL Sharp, Bevel, Curved, Non-coring 0.15 mm 0.72 mm Each
10303621 500 μL Sharp, Bevel, Curved, Non-coring 0.41 mm 0.72 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Product Code. 10313571 Supplier Hamilton™ Supplier No. 80600
  • ONLINE EXCLUSIVE PRICE
    1040.00 NOK
    781.67 NOK / Pack of 1
    Save 258.33 NOK
    24% Off
Estimated Shipment: 22-12-2026
The 700 series syringe is the original hand-fitted Hamilton syringe. This series of syringes is ideal for dispensing volumes from 0.5 μL up to 500 μL. A variety of different terminations and needle options are available in this series.
  • Original hand-fitted Hamilton syringe
  • Ideal for dispensing volumes from 0.5 µL up to 500 µL
  • A variety of different terminations and needle options are available in this series
Specifications
Autoclavable Not Autoclavable
Material Stainless Steel (Plunger)
Needle Gauge 22s G
Diameter (English) Inner 0.061 in.
Diameter (English) Inner Needle 0.0059 in.
Diameter (Metric) Inner Needle 0.15 mm
Diameter (English) Outer Needle 0.0283 in.
Diameter (English) Outer 0.305 in.
Diameter (Metric) Inner 1.46 mm
Diameter (Metric) Outer 7.75 mm
Diameter (Metric) Outer Needle 0.72 mm
For Use With (Application) For dispensing low volumes to achieve the highest level of accuracy and precision possible
Length (English) 2 in.
Length (English) Needle 2 in.
Length (Metric) 51 mm
Product Type Microliter Syringe
Max. Pressure 2000 psi
Max. Temperature (English) 122°F
Max. Temperature (Metric) 50°C
Min. Temperature (English) 50°F
Model 710
Needle Point Number 2
Quantity Ea.
Volume (Metric) 100 μL
Needle Point Style Sharp, Bevel, Curved, Non-coring
Connection Type Cemented