Description
- Made from high-tech Kevlar™ Aramid fibers para-aramid short fibers
- Neptune coating allows glove fibers to repel liquids
- Seamless construction to avoid irritation
- Short fibers provide extra comfort
- Terry-loop technique enchances heat insulation
- Thick wall offers high shock absorption
- Ambidextrous
- Washable and reusable
Specifications
Specifications
| Back Material | Neptune™ |
| Certifications/Compliance | Standards CATEGORY III EN 388 : 254X, EN 407, ANSI |
| Cuff Material | Neptune™ |
| Coating Material | Uncoated |
| Color | Yellow |
| Cut Resistance Rating | ANSI A2 |
| Hand | Specific |
| Latex Free | Yes |
| Material | Aramid Fiber |
| Product Type | Heavyweight Gloves |
| Show More |