missing translation for 'onlineSavingsMsg'
Learn More

7-Nitroindole-2-carboxylic acid, 96%

Product Code. p-7025353
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11435686 1 g 1g
11445686 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11435686

Brand: Thermo Scientific Alfa Aesar B20786.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6960-45-8 | C9H6N2O4 | 206.157 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6960-45-8
Molecular Formula C9H6N2O4
Molecular Weight (g/mol) 206.157
MDL Number MFCD00044720
InChI Key BIUCOFQROHIAEO-UHFFFAOYSA-N
Synonym 7-nitro-1h-indole-2-carboxylic acid, 7-nitroindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 7-nitro, 7-nitroindole-2-carboxylicacid, dna base excision repair pathway inhibitor, maybridge1_004786, indole-2-carboxylic acid, 7-nitro, pubchem9857, acmc-1b8pb, 1h-indole-2-carboxylicacid, 7-nitro
PubChem CID 81409
SMILES C1=CC2=C(C(=C1)[N+](=O)[O-])NC(=C2)C(=O)O

Specifications

Melting Point 264°C to 266°C
Quantity 1 g
Formula Weight 206.16
Percent Purity 96%
Chemical Name or Material 7-Nitroindole-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.