Learn More
7-Methylxanthine, 98%
Product Code. 11412260
Change view
Click to view available options
Quantity:
50 mg
250 mg
Unit Size:
250mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412260 50 mg 50mg
15433847 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11412260 Supplier Thermo Scientific Alfa Aesar Supplier No. H29190.MF
  • ONLINE EXCLUSIVE PRICE
    1290.00 NOK
    1001.65 NOK / 50mg
    Save 288.35 NOK
    22% Off
 Restricted Item
In Stock
Add to basket
CAS: 552-62-5 | C6H6N4O2 | 166.14 g/mol
It is used to metabolite theophylline and caffeine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used to metabolite theophylline and caffeine.
Solubility
Soluble in dimethyl sulfoxide. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 552-62-5
Molecular Formula C6H6N4O2
Molecular Weight (g/mol) 166.14
MDL Number MFCD00037979
InChI Key PFWLFWPASULGAN-UHFFFAOYSA-N
Synonym 7-methylxanthine, heteroxanthine, heteroxanthin, 7-methylxanthin, 7-methyl-1h-purine-2,6 3h,7h-dione, 2,6-dihydroxy-7-methylpurine, xanthine, 7-methyl, 3,7-dihydro-7-methyl-1h-purine-2,6-dione, 7-methyl-3,7-dihydro-1h-purine-2,6-dione, 1h-purine-2,6-dione, 3,7-dihydro-7-methyl
PubChem CID 68374
ChEBI CHEBI:48991
SMILES CN1C=NC2=C1C(=O)NC(=O)N2
Specifications
Melting Point >300°C
Quantity 50 mg
Solubility Information Soluble in dimethyl sulfoxide. Insoluble in water.
IUPAC Name 7-methyl-3H-purine-2,6-dione
Formula Weight 166.14
Percent Purity 98%
Chemical Name or Material 7-Methylxanthine