Learn More
7-Methylindole, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 7-Methylindole |
| CAS | 933-67-5 |
| Molecular Formula | C9H9N |
| Quantity | 5 g |
| Synonym | 7-Methyl-1H-indole, NSC 618 |
| InChI Formula | InChI=1S/C9H9N/c1-7-3-2-4-8-5-6-10-9(7)8/h2-6,10H,1H3 |
| SMILES | Cc1cccc2cc[nH]c12 |
| IUPAC Name | 7-methyl-1H-indole |
| Molecular Weight (g/mol) | 131.17 |
| Formula Weight | 131.0735 |