Learn More
7-Methylguanosine, MedChemExpress
Product Code. 16401287
Encompass_Preferred
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Product Code. 16401287
Brand: Medchem Express HY122524
MedChemExpress 7-Methylguanosine is a novel cNIIIB nucleotidase inhibitor with IC50 value of 87.8 ± 7.5 μM.
7-Methylguanosine is a novel cNIIIB nucleotidase inhibitor with IC50 value of 87.8 ± 7.5 μM.
Specifications
Chemical Name or Material 7-Methylguanosine
CAS 20244-86-4
Color White
Purity Grade Notes Research
Molecular Formula C11H15N5O5
Quantity 50 mg
Solubility Information H2O : 25 mg/mL (84.10 mM; ultrasonic and warming and heat to 60°C) ∣DMSO : 12.5 mg/mL (42.05 mM; Need ultrasonic)
SMILES [O-]C1=NC(N)=NC2=C1[N+](C)=CN2[C@H]3[C@@H]([C@@H]([C@H](O3)CO)O)O
Molecular Weight (g/mol) 297.27
Formula Weight 297.27
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.