Learn More
7-Methoxy-1-naphthaleneacetonitrile, Mikromol™
Description
7-Methoxy-1-naphthaleneacetonitrile
Specifications
Specifications
| Chemical Name or Material | 7-Methoxy-1-naphthylacetonitrile |
| CAS | 138113-08-3 |
| Analyte or Component Names | 7-Methoxy-1-naphthylacetonitrile |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H11 N O |
| Certifications/Compliance | ISO 17025 |
| Synonym | 7-Methoxy-1-naphthaleneacetonitrile, (7-Methoxynaphthalen-1-yl)acetonitrile, 1-Cyanomethyl-7-methoxynaphthalene, 2-(7-Methoxy-1-naphthyl)acetonitrile, 2-(7-Methoxynaphthalen-1-yl)acetonitrile, 7-Methoxy-1-naphthylacetonitrile, 7-Methoxynaphthalene-1-acetonitrile |
| InChI Formula | InChI=1S/C13H11NO/c1-15-12-6-5-10-3-2-4-11(7-8-14)13(10)9-12/h2-6,9H,7H2,1H3 |
| SMILES | COc1ccc2cccc(CC#N)c2c1 |
| IUPAC Name | 2-(7-methoxynaphthalen-1-yl)acetonitrile |
| Show More |