Learn More
7-Hydroxy-3,4-dihydro-2(1H)-quinolinone, 97%, Thermo Scientific™
Product Code. 15423697
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15423697 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15423697 Supplier Thermo Scientific Alfa Aesar Supplier No. H27485.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
CAS 22246-18-0
Melting Point 230°C to 234°C
Molecular Formula C9H9NO2
MDL Number MFCD06410891
Quantity 1 g
Synonym 3,4-dihydro-7-hydroxy-2 1h-quinolinone, 7-hydroxy-3,4-dihydroquinolin-2 1h-one, 7-hydroxy-3,4-dihydro-2 1h-quinolinone, 3,4-dihydro-7-hydroxycarbostyril, 7-hydroxy-3,4-dihydro-quinolin-2-one, 7-hydroxy-3,4-dihydrocarbostyril, 7-hydroxy-3,4-dihydro carbostyril, 7-hydroxy-1,2,3,4-tetrahydroquinolin-2-one, 2 1h-quinolinone, 3,4-dihydro-7-hydroxy, 7-hydroxy-2-oxo-1,2,3,4-tetrahydroquinoline
InChI Key LKLSFDWYIBUGNT-UHFFFAOYSA-N
SMILES C1CC(=O)NC2=C1C=CC(=C2)O
IUPAC Name 7-hydroxy-3,4-dihydro-1H-quinolin-2-one
Molecular Weight (g/mol) 163.176
PubChem CID 2785758
Formula Weight 163.17
Percent Purity 97%
Chemical Name or Material 7-Hydroxy-3,4-dihydro-2(1H)-quinolinone