Learn More
7-Fluoro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 97%
Product Code. 15259686 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15259686 1 g 1g
15259656 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15259686 Supplier Thermo Scientific Acros Supplier No. 450460010
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 799274-06-9 | C9H10FN·ClH | 187.64 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 799274-06-9
Molecular Formula C9H10FN·ClH
Molecular Weight (g/mol) 187.64
InChI Key LOJNQXIJLCPQDR-UHFFFAOYSA-N
Synonym 7-fluoro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 7-fluoro-1,2,3,4-tetrahydro-isoquinoline hydrochloride, 7-fluoro-1,2,3,4-tetrahydroisoquinoline hcl, 7-fluoro-1,2,3,4-tetrahydroisoquinolinehydrochloride, 7-fluoro-1,2,3,4-tetrahydroisoquinoline hydrochloride salt, isoquinoline, 7-fluoro-1,2,3,4-tetrahydro-, hydrochloride, 7-fluoro-1,2,3,4-tetrahydro-isoquinoline hydrochloride salt, 7-fluoro-1,2,3,4-tetrahydroisoquinoline-hydrogen chloride 1/1
PubChem CID 45073971
IUPAC Name 7-fluoro-1,2,3,4-tetrahydroisoquinoline;hydrochloride
SMILES C1CNCC2=C1C=CC(=C2)F.Cl
Specifications
Melting Point 177.0°C to 179.0°C
Infrared Spectrum Authentic
Assay Percent Range 96.0 to max. 104% min. (Argentometry)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 187.64
Percent Purity 97%
Chemical Name or Material 7-Fluoro-1,2,3,4-tetrahydroisoquinoline hydrochloride
compliance-icons
Product Identifier
  • 7-Fluoro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.