Learn More
7-Chloroquinoline, 97%, Thermo Scientific™
Product Code. 15555916 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15555916 5 g 5g
15545916 1 g 1g
2 options
Product Code. 15555916
Brand: Thermo Scientific Acros 456010050
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 612-61-3
Molecular Formula C9H6ClN
Molecular Weight (g/mol) 163.61
InChI Key QNGUPQRODVPRDC-UHFFFAOYSA-N
Synonym quinoline, 7-chloro, 7-chloro-quinoline, pubchem19899
PubChem CID 521963
IUPAC Name 7-chloroquinoline
SMILES C1=CC2=C(C=C(C=C2)Cl)N=C1
Specifications
Assay Percent Range 97%
Quantity 5 g
Formula Weight 163.61
Percent Purity 97%
Chemical Name or Material 7-Chloroquinoline
compliance-icons
Product Identifier
  • 7-Chloroquinoline, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.