Learn More
7-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid hydrate, 95%, Thermo Scientific™
Product Code. 15428975
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Product Code. Quantity unitSize
15428975 250 mg 250mg
1 options
Product Code. 15428975
Brand: Thermo Scientific Alfa Aesar H35003.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
CAS 1019018-46-2
Melting Point 234°C to 235°C
Molecular Formula C8H5BrN2O2
MDL Number MFCD09995206
Quantity 250 mg
Synonym 7-bromoimidazo 1,2-a pyridine-2-carboxylic acid, 7-bromo-imidazo 1,2-a pyridine-2-carboxylic acid, 7-bromoh-imidazo 1,2-a pyridine-2-carboxylic acid, 7-bromoimidazo 1,2-a pyridine-2-carboxylic acid hydrate, 7-bromoimidazo 1,2-a pyridine-2-carboxylicacid, 7-?bromoimidazo 1,?2-?a ?pyridine-?2-?carboxylic a, 7-bromo-4-hydroimidazo 1,2-a pyridine-2-carboxylic acid
InChI Key HKOPWQHBKVSTHO-UHFFFAOYSA-N
SMILES OC(=O)C1=CN2C=CC(Br)=CC2=N1
IUPAC Name 7-bromoimidazo[1,2-a]pyridine-2-carboxylic acid
Molecular Weight (g/mol) 241.04
PubChem CID 51063866
Formula Weight 241.05 (Anhydrous)
Percent Purity 95%
Chemical Name or Material 7-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid hydrate